Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXQ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540772
Description
Peptide sequence: MKLEVFVPRA AHGDKQGSDL EGAGGSDAPS PLSAAGDDSL GSDGDCAANS Sequence homology: Human: 100%; Mouse: 85%; Pig: 86%; Rat: 85%.
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
Specifications
FOXQ1 | |
Polyclonal | |
Unconjugated | |
FOXQ1 | |
forkhead box protein Q1; forkhead box Q1; FOXQ1; Hepatocyte nuclear factor 3 forkhead homolog 1; Hfh1; HFH-1; Hfh1l; HFH-1l; HNF-3/forkhead homolog 1 like; HNF-3/forkhead homolog-1; HNF-3/forkhead-like protein 1; sa; satin; winged helix/forkhead transcription factor | |
Rabbit | |
Affinity chromatography | |
RUO | |
94234 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9C009 | |
FOXQ1 | |
Synthetic peptide directed towards the N-terminal of human FOXQ1 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction