Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536475 Supplier Invitrogen™ Supplier No. PA536475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 35 kDa. Purity is >95% by SDS-PAGE.

The forkhead-box (FOX) genes comprise a superfamily of at least 43 members that express proteins which are involved in transcriptional regulation and may be associated with the pathogenesis of various cancers. FOX family members are monomeric, helix-turn-helix proteins with a core DNA-binding domain of approximately 110 amino acids. FOX transcription factors play roles in determining cell fate during early development. FOXR1 (forkhead box protein R1), also known as FOXN5 (forkhead box protein N5) or DLNB13, is a 292 amino acid nuclear protein that belongs to the FOX family and contains one forkhead DNA-binding domain. Through chromosomal aberrations such as retroviral integration, gene amplification or translocation, FOXR1 may be involved the development of certain invasive carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXR1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene Foxr1
Gene Accession No. Q3UTB7, Q6PIV2
Gene Alias DLNB13; Forkhead box protein N5; forkhead box protein R1; forkhead box R1; forkhead box R1 variant 1; forkhead box R1 variant 2; Foxn5; FOXR1; Gm1115
Gene Symbols Foxr1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 232-286 of human FoxR1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283150, 315601, 382074
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.