Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXR2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550223

Catalog No. PA550223


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total FOXR2 protein.

Forkhead-box (FOX) transcription factors are implicated in carcinogenesis through gene amplification, retroviral integration, or chromosomal translocation. Human FOXN6 mRNA was expressed in breast cancer cell line and primary breast cancer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXR2
Polyclonal
Unconjugated
FOXR2
EG436240; Forkhead box protein N6; forkhead box protein R2; forkhead box R2; Foxn6; Foxr2
Rabbit
Antigen Affinity Chromatography
RUO
139628
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6PJQ5
FOXR2
Full length fusion protein of Human FOXR2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.