Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584337 Supplier Invitrogen™ Supplier No. PA584337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CEFWYSLHGQ VPGLLDWDMR NELFLPCTTD QCSLAEQILA KYRVGVMKPP EMPQKRRPSP DGDGPPCEPN LWM Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 68%.

Forkhead-box (FOX) transcription factors are implicated in carcinogenesis through gene amplification, retroviral integration, or chromosomal translocation. Human FOXN6 mRNA was expressed in breast cancer cell line and primary breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXR2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOXR2
Gene Accession No. Q6PJQ5
Gene Alias EG436240; Forkhead box protein N6; forkhead box protein R2; forkhead box R2; Foxn6; Foxr2
Gene Symbols FOXR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FOXR2. Recombinant protein control fragment (Product #RP-105069).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139628
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.