Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXRED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100532 Supplier Invitrogen™ Supplier No. PA5100532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FOXRED1.

This gene encodes a protein that contains a FAD-dependent oxidoreductase domain. The encoded protein is localized to the mitochondria and may function as a chaperone protein required for the function of mitochondrial complex I. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXRED1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FOXRED1
Gene Accession No. Q3TQB2, Q96CU9
Gene Alias BC024806; FAD dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain-containing protein 1; Foxred1; FP634; H17; RGD1311785; TEG-23; testis expressed gene 23; Tex23
Gene Symbols FOXRED1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FOXRED1(Accession Q96CU9), corresponding to amino acid residues G334-Q384.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235169, 315547, 55572
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.