Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXRED1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561178
Description
Immunogen sequence: LSVGGIRQQF SLPENIQLSL FSASFLRNIN EYLAVVDAPP LDLRFNPSGY LLLASEKDAA AMESNVKVQR QEGAKVSLMS PDQLRNK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
This gene encodes a protein that contains a FAD-dependent oxidoreductase domain. The encoded protein is localized to the mitochondria and may function as a chaperone protein required for the function of mitochondrial complex I. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants have been observed for this gene.
Specifications
FOXRED1 | |
Polyclonal | |
Unconjugated | |
FOXRED1 | |
BC024806; FAD dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain-containing protein 1; Foxred1; FP634; H17; RGD1311785; TEG-23; testis expressed gene 23; Tex23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CU9 | |
FOXRED1 | |
Recombinant protein corresponding to Human FOXRED1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction