Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXRED2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556797 Supplier Invitrogen™ Supplier No. PA556797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVLGAGPAGL QMAYFLQRAG RDYAVFERAP RPGSFFTRYP RHRKLISINK RYTGKANAEF NLRHDWNSLL SHDPRLLFRH YSRAYFPDAR DMVRYLGDFA DTLGLRVQYN TTIAHVTLDK DRQAWNGHYF ILTDQKG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

FOXRED2, also known as ERFAD, is an endoplasmic reticulum (ER) luminal flavoprotein which may function in ER associated degradation (ERAD). It has been proposed to bind non-native proteins in the endoplasmic reticulum and target them to the ubiquitination machinery for subsequent degradation. Upon knockdown of FOXRED2, the degradation of the ERAD model substrate ribophorin 332 is delayed, and the overall level of polyubiquitinated cellular proteins is decreased. Defects in the related protein FOXRED1 are a cause of mitochondrial complex I deficiency (MT-C1D), suggesting the FOXRED2 may also play a role in MT-C1D.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXRED2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOXRED2
Gene Accession No. Q8IWF2
Gene Alias Endoplasmic reticulum flavoprotein associated with degradation; ER flavoprotein associated with degradation; ERFAD; FAD dependent oxidoreductase domain containing 2; FAD-dependent oxidoreductase domain containing 2; FAD-dependent oxidoreductase domain-containing protein 2; FOXRED2
Gene Symbols FOXRED2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FOXRED2. Recombinant protein control fragment (Product #RP-91545).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.