Learn More
Invitrogen™ FOXS1 Polyclonal Antibody

Description
The antibody detects endogenous level of total FOXS1 protein.
Specifications
Specifications
| Antigen | FOXS1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | FOXS1 |
| Gene Accession No. | O43638, Q61574 |
| Gene Alias | Fkh3; Fkhl18; forkhead box protein S1; forkhead box S1; forkhead-like 18; forkhead-like 18 protein; forkhead-related activator 10; forkhead-related transcription factor 10; forkhead-related transcription factor FREAC-10; Foxs1; FREAC10; FREAC-10; Transcription factor FKH-3 |
| Gene Symbols | FOXS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.