Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FPR1/2 Monoclonal Antibody (NFPR1), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 30107100UG

This item is not returnable.
View return policy
Description
FPR1/2 Monoclonal antibody specifically detects FPR1/2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
FPR1/2 | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P21462, P25090 | |
His-tagged recombinant FPR1 (human neutrophil N-formyl peptide receptor 1) expressed in and purified from Sf9 cells. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
NFPR1 | |
Unconjugated | |
FPR1 | |
Mouse | |
Protein A | |
RUO | |
2357, 2358 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction