Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR1 Antibody (350418), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB371546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371546 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371546 Supplier Novus Biologicals Supplier No. FAB3744E
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FPR1 Monoclonal antibody specifically detects FPR1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen FPR1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 350418
Conjugate DyLight 405
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias fMet-Leu-Phe receptor, FMLP, fMLP receptor, formyl peptide receptor 1, FPRfMet-Leu-Phe receptor, N-formyl peptide receptor, N-formylpeptide chemoattractant receptor
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FPR1, Met1-Lys350, Accession # NP_002020
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2357
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.