Learn More
FPR1 Polyclonal Antibody

Description
FPR1 Polyclonal Antibody for Western Blot, IHC
Specifications
Specifications
| Antigen | FPR1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 |
| Gene | FPR1 |
| Gene Accession No. | P21462 |
| Gene Alias | fMet-Leu-Phe receptor; fMLF-R; FMLP; fMLP receptor; formyl peptide receptor 1; FPR; Fpr receptor; FPR1; FPR-1; lipoxin A4 receptor; LXA4R; N-formyl peptide receptor; N-formylpeptide chemoattractant receptor |
| Gene Symbols | FPR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.