Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPR2 Polyclonal Antibody, Huabio

Catalog No. 502544396
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4396 Supplier HUABIO RESEARCH INC Supplier No. R151013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FPR2 Polyclonal antibody specifically detects FPR2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FPR2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene FPR2
Gene Accession No. O88536, P25090
Gene Alias ALXR; E330010I07Rik; fMLPR1; FMLP-related receptor I; FMLP-R-I; FMLP-R-II; FMLPX; formyl peptide receptor 2; formyl peptide receptor, related sequence 2; formyl peptide receptor, related sequence 2-like; formyl peptide receptor-like 1; Formylpeptide receptor-related sequence 2; FPR2; FPR2A; FPRH1; FPRH2; Fprl1; Fprl-1; FPR-like 1; Fpr-rs1; Fpr-rs2; HM63; Lipoxin A4 receptor; lipoxin A4 receptor (formyl peptide receptor related); lipoxin A4 receptor-like protein; Lipoxin receptor; LXA4 receptor; LXA4R; N-formyl peptide receptor 2; N-formylpeptide receptor-like 2; RFP
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal mouse FPR2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14289, 2358, 690158
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.