Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FPR3 Monoclonal Antibody (8A6G12)
GREENER_CHOICE

Catalog No. FERMA548460
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548460 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548460 Supplier Invitrogen™ Supplier No. MA548460
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FPR3 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

The N-formyl peptide receptor (FPR) family is comprised of three members, FPR, FPR3 (also designated FPRL1, lipoxin A4 receptor, FPRH1 or FPR2) and FPR like-2 (FPRL2), all of which are chemotactic G protein-coupled receptors that contain seven transmembrane domains. These receptors are found on the surface of phagocytic leukocytes, such as neutrophils and monocytes, and each family member contains specific residues, which are responsible for determining its ligand specificity. FPR3 is a promiscuous receptor that binds to several ligands, including lipoxin A4, N-formyl-methionyl-leucyl-phenylalanine (fMLP), serum amyloid A (SAA), prion peptide and the 42 amino acid form of beta amyloid. Upon activation, FPR3 induces migration and calcium mobilization in human monocytes and neutrophils and is involved in inflammatory and host defense responses. FPR3 may mediate inflammation in prion and Alzheimer's diseases, which makes it a potential target for therapeutic agents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FPR3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 8A6G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene FPR3
Gene Accession No. P25089
Gene Alias ALX; FML2_HUMAN; FMLP-related receptor I; FMLP-related receptor II; FMLP-R-I; FMLP-R-II; FMLPY; formyl peptide receptor 3; Formyl peptide receptor related sequence 1; Formyl peptide receptor-like 1; formyl peptide receptor-like 2; Formyl peptide receptor-related sequence 1; Fpr2; FPR3; FPRH1; FPRH2; Fprl1; FPRL2; Fpr-rs1; Fpr-s1; Lipoxin A4 receptor; LXA4 receptor; Lxa4r; LXA4-R; N-formyl peptide receptor 2; N-formyl peptide receptor 3; N-formylpeptide receptor-like 1; RGD1566044; RMLP-R-I
Gene Symbols FPR3
Host Species Mouse
Immunogen Purified recombinant fragment of human FPR3 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.