Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPRL1/Lipoxin A4 receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985559
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5559 Supplier Bioss Supplier No. BS3654R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Formyl peptide receptor 2 (FPR2) is a low affinity receptor for N-formyl-methionyl peptides. Binding of FMLP to FPR2 causes activation of neutrophils. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. The activation of LXA4R could result in an anti-inflammatory outcome counteracting the actions of proinflammatory signals such as LTB4.

Specifications

Antigen FPRL1/Lipoxin A4 receptor
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FPR2
Gene Accession No. P25090
Gene Alias ALXR; E330010I07Rik; fMLPR1; FMLP-related receptor I; FMLP-R-I; FMLP-R-II; FMLPX; Formyl peptide receptor 2; formyl peptide receptor, related sequence 2; formyl peptide receptor, related sequence 2-like; Formyl peptide receptor-like 1; Formylpeptide receptor-related sequence 2; FPR2; FPR2A; FPRH1; FPRH2; FPRL1; Fprl-1; FPR-like 1; Fpr-rs1; Fpr-rs2; HM63; Lipoxin A4 receptor; lipoxin A4 receptor (formyl peptide receptor related); Lipoxin A4 receptor-like protein; Lipoxin receptor; LXA4 receptor; LXA4R; N-formyl peptide receptor 2; N-formylpeptide receptor-like 2; RFP
Gene Symbols FPR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FPRL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2358
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.