Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FPRL2/FPR3 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3896U100UG
This item is not returnable.
View return policy
Description
FPRL2/FPR3 Monoclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry.Specifications
FPRL2/FPR3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P25089 | |
FPR3 | |
HEK293 human embryonic kidney cell line transfected with human FPRL2 Met1-Met352 Accession # P25089 | |
RUO | |
2359 | |
Human | |
IgG2b |
Flow Cytometry | |
374822 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPR3, FPRH1, FPRH1FPRH2, FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I | |
Mouse | |
100 μg | |
Primary | |
Detects human FPRL2. Stains human FPRL2 transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction