Learn More
International Enviroguard™ FR and Chemical Resistant Coverall with Attached Hood and Elastic Wrists
Catalog No. 17184020 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
PyroGuard CRFR flame-resistant coveralls are designed to wear over your primary FR garments, helping to extend the life of your primary FR clothing.
  • Coveralls also offer protection from a broad range of chemicals and provide flash fire protection that meets NFPA 2113
  • Advanced, self-extinguishing fabric does not melt or drip when ignited to prevent flames from spreading
  • Attached respirator-fit hood combined with secure, elastic wrists accommodate a respirator to prevent toxic inhalation and keep sleeves in place to avoid skin exposure
  • PyroGuard CRFR flame-resistant suits stop flames and chemicals in their tracks offering the comfort and safety you need to work in tough environments
  • Proprietary bilaminate fabric protects against flames and chemicals
  • Elastic back stretches with user for more comfort and better tear resistance
  • Storm flap over zipper adds an extra layer of protection
  • Taped seams offer added protection over serged seam coveralls
  • Possible applications include petrochemical environments, manufacturing, industrial, oil/gas, and more
Specifications
Product Type Flame-resistant Coveralls
Size 2X-Large
Gender Unisex
Material CFR Bilaminate Fabric
Color Gray
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Open
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Acid, Chemicals, Flame, Caustics, Anti-static
Latex Free Yes
No. of Pockets None
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.