Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Women's FR/CP Lab Coats

Catalog No. 19800072 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-800-072 3X-Large
19-800-070 X-Small
19-800-068 Small
19-800-067 Medium
19-800-066 Large
19-800-069 X-Large
19-800-071 2X-Large
19-800-073 4X-Large
19-800-074 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-800-072 Supplier Bulwark™ Supplier No. KNR3RB 3L 00
This item is not returnable. View return policy

Lightweight and breathable lab coat provides dual hazard protection with flame resistance and chemical protection.

  • Nomex™ IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
  • ShieldCXP repels small amounts of liquid chemicals that are either dropped or splashed on the fabric
  • Nomex™ knit cuffs are fire resistant but do not provide chemical protection - these are typically covered by gloves
  • Black lay-down collar
  • Concealed snap front closure
  • Splash-resistant side pass-through for easy inner pocket access
  • Pockets include one double-stitched chest pocket and two large double-stitched lower front pockets with snap flaps
  • Available in a variety of sizes
  • Includes a waistband in the back for a more tailored fit

Certifications

NFPA 2112 UL-Certified

Compliance

Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire. UL certified.

Specifications

Product Type Flame-resistant Lab Coat
Size 3X-Large
Gender Women's
Material Nomex IIIA
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Style Regular
No. of Pockets 3
Finish Westex ShieldCXP Chemical Splash Protection
Model 354CH45RB
Size (English) Chest 49.5 in.
Size (Metric) Chest 125.7 cm
Length (English) Sleeve 35.5 in.
Length (Metric) Sleeve 90.2 cm
Weight (English) 4.5 oz.
Weight (Metric) 128 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.