Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FR4 (Folate receptor 4) Rat anti-Mouse, Brilliant Violet 786, Clone: 12A5, BD Optibuild™

Catalog No. BDB744124 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744124 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744124 Supplier BD Biosciences Supplier No. 744124
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibodies TH6 and 12A5 recognize Folate Receptor 4(FR4), also known as the membrane folate- binding protein 3(FBP3). FR4 is a heavily glycosylated 35 kD receptor expressed exclusively in lymphoid tissue and an isoform of the family of receptors that recognize the essential nutrient folic acid. Natural T regulatory cells constitutively express high levels of FR4. Differential expression of FR4 in combination with CD25 can distinguish four functionally distinct CD4+ T cell subpopulations; Natural Tregs, effector T cells, memory-like T cells and Naοve T cells. FR4hi CD25+ expressing CD4+ T cells also express high amounts of Foxp3, GITR and CTLA-4.Monoclonal antibody TH6 and 12A5 stained CD25+CD4+ T cells at a higher level than other CD4+ or CD8+ T cells. In addition, in vivo injection of TH6 monoclonal antibody reduced the number of CD25+CD4+ T cells and CD25-CD4+ T cells in peripheral blood. Clone 12A5 has been demonstrated to work in western blot. Clones TH6 and 12A5 do not block one another in a flow cytometric assay.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FR4 (Folate receptor 4)
Classification Monoclonal
Clone 12A5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Folate receptor 4, FBP, FBP3, FRdelta, FRd
Host Species Rat
Immunogen Mouse CD4+ CD25+ T cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.