Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FR4 (Folate receptor 4) Rat anti-Mouse, BUV395, Clone: 12A5, BD Optibuild™

Catalog No. BDB744125 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744125 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744125 Supplier BD Biosciences Supplier No. 744125
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibodies TH6 and 12A5 recognize Folate Receptor 4(FR4), also known as the membrane folate- binding protein 3(FBP3). FR4 is a heavily glycosylated 35 kD receptor expressed exclusively in lymphoid tissue and an isoform of the family of receptors that recognize the essential nutrient folic acid. Natural T regulatory cells constitutively express high levels of FR4. Differential expression of FR4 in combination with CD25 can distinguish four functionally distinct CD4+ T cell subpopulations; Natural Tregs, effector T cells, memory-like T cells and Naοve T cells. FR4hi CD25+ expressing CD4+ T cells also express high amounts of Foxp3, GITR and CTLA-4.Monoclonal antibody TH6 and 12A5 stained CD25+CD4+ T cells at a higher level than other CD4+ or CD8+ T cells. In addition, in vivo injection of TH6 monoclonal antibody reduced the number of CD25+CD4+ T cells and CD25-CD4+ T cells in peripheral blood. Clone 12A5 has been demonstrated to work in western blot. Clones TH6 and 12A5 do not block one another in a flow cytometric assay.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FR4 (Folate receptor 4)
Classification Monoclonal
Clone 12A5
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Folate receptor 4, FBP, FBP3, FRdelta, FRd
Host Species Rat
Immunogen Mouse CD4+ CD25+ T cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.