Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FR4 Monoclonal Antibody (eBio12A5), FITC, eBioscience™, Invitrogen™

Catalog No. 509835
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-983-5 25 μg
50-983-6 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-983-5 Supplier Invitrogen™ Supplier No. 11544580
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody eBio12A5 recognizes FR4, also known as Folate receptor 4, FR delta and FBP3 (folate binding protein3). FR4 is a heavily glycosylated, 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural Treg have high levels of FR4 and together with CD25 can be used to distinguish natural Treg, effector T cells, memory-like T cells and naive T cells. FR4 high CD25 high cells are natural Treg with high levels of Foxp3. FR4 high CD25 low cells are identified as central memory T cells, which upon stimulation, can proliferate and produce large amounts of IL-2. In contrast, FR4 low CD25 high cells secrete proinflammatory cytokines such as IFN gamma and IL-17 and have been characterized as effector memory T cells. Based on co-staining studies, the epitopes recognized by eBioTH6 and eBio12A5 are different, thereby allowing functional studies with eBioTH6 to be evaluated with eBio12A5. Applications Reported: This eBio12A5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio12A5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

FR4 is a heavily glycosylated 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural T regs have high levels of FR4 and together with CD25 can be used to distinguish natural Tregs, effector T cells, memory-like T cells and naive T cells. In vivo studies with eBioTH6 show a decrease in the number of CD4+CD25+ cells and an enhanced tumor response leading to the elimination of advanced stages of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FR4
Applications Flow Cytometry
Classification Monoclonal
Clone eBio12A5
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Izumo1r
Gene Accession No. Q9EQF4
Gene Alias 0910001L11Rik; C86255; Folate receptor 4; folate receptor 4 (delta); folate receptor 4 (delta) homolog; folate receptor 4, delta (putative); folate receptor delta; Folate-binding protein 3; Folbp3; Folr4; FR-delta; IZUMO1 receptor protein JUNO; IZUMO1 receptor, JUNO; Izumo1r; Juno; probable folate receptor delta; sperm-egg fusion protein Juno
Gene Symbols Izumo1r
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64931
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.