Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRA-1/FOSL1 Antibody (OTI12F9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271351F
Description
FRA-1/FOSL1 Monoclonal specifically detects FRA-1/FOSL1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
FRA-1/FOSL1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FOSL1 | |
Full-length protein expressed in 293T cell transfected with human FRA-1/FOSL1 expression vector. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunohistochemistry | |
OTI12F9 | |
Immunohistochemistry | |
FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, fra-1, FRA1 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Wnt Signaling Pathway | |
8061.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction