Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRA-1/FOSL1 Antibody (OTI12F9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147757
Description
FRA-1/FOSL1 Monoclonal specifically detects FRA-1/FOSL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FRA-1/FOSL1 | |
| Monoclonal | |
| 1.1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50 | |
| FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, fra-1, FRA1 | |
| Mouse | |
| 29.4 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 8061 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI12F9 | |
| Unconjugated | |
| P15407 | |
| FOSL1 | |
| Full-length protein expressed in 293T cell transfected with human FOSL1 expression vector | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens FOS-like antigen 1 (FOSL1). | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction