Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRA-1/FOSL1 Antibody (SR1652), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322276100UL
Description
FRA-1/FOSL1 Monoclonal antibody specifically detects FRA-1/FOSL1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
FRA-1/FOSL1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
SR1652 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, FRA1, FRA-1 | |
A synthesized peptide derived from human FRA-1/FOSL1 (Uniprot #: P15407) | |
100 μg | |
Cell Cycle and Replication, Wnt Signaling Pathway | |
8061 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction