Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fra1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566880 Supplier Invitrogen™ Supplier No. PA566880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVPSINTMSGS QELQWMVQPH FLGPSSYPRP LTYPQYSPPQ PRPGVIRALG PPPGVRRRPC EQISPE Highest antigen sequence identity to the following orthologs - mouse 88%, rat 96%.

The v-Fos oncogene was initially detected in two independent murine osteosarcoma virus isolates and an avian nephroblastoma virus. Members of the c-Fos gene family, including c-Fos, Fos B, Fra-1 and Fra-2, encode nuclear phosphoproteins that are rapidly and transiently induced by a variety of agents and function as transcriptional regulators for several genes. In contrast to c-Jun proteins, which form homo- and heterodimers that bind to specific DNA response elements, c-Fos proteins are only active as heterodimers with members of the Jun gene family. In addition, selected ATF/CREB family members can form leucine zipper dimers with Fos and Jun. Different dimers exhibit differential specificity and affinity for AP-1 and CRE sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fra1
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOSL1
Gene Accession No. P15407
Gene Alias AW538199; FOS like 1, AP-1 trancription factor subunit; FOS like 1, AP-1 transcription factor subunit; FOS like antigen 1; Fosl1; FOS-like antigen 1; FOS-like antigen-1; fos-related antigen 1; FRA; Fra1; fra-1; HGNC:13718; LOW QUALITY PROTEIN: fos-related antigen 1
Gene Symbols FOSL1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human FOSL1 (Product #RP-107529)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.