Learn More
FRA10AC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304738AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a nuclear phosphoprotein of unknown function. The 5' UTR of this gene is part of a CpG island and contains a tandem CGG repeat region that normally consists of 8-14 repeats but can expand to over 200 repeats. The expanded allele becomes hypermethylated and is not transcribed; however, an expanded repeat region has not been associated with any disease phenotype. This gene is found within the rare FRA10A folate-sensitive fragile site.Specifications
FRA10AC1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q70Z53, Q8BP78 | |
FRA10AC1 | |
Between 265 and 315 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FRA10AC1 | |
C10orf4, F26C11.1-like, FRA10A, fragile site 10q23.3, protein FRA10AC1, rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118924, 70567 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.