Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRA2 Antibody (PCRP-FOSL2-1B1), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308467DL594
Description
FRA2 Monoclonal antibody specifically detects FRA2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
FRA2 | |
Monoclonal | |
DyLight 594 | |
FOS-like antigen 2, fos-related antigen 2, FRA-2, FRA2FLJ23306 | |
Recombinant full-length human FRA2 protein (Uniprot: P15408) | |
100 μg | |
Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
2355 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, ELISA, Immunofluorescence, Protein Array | |
PCRP-FOSL2-1B1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction