Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fra2 Monoclonal Antibody (OTI2B4)
GREENER_CHOICE

Catalog No. PIMA527192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527192 Supplier Invitrogen™ Supplier No. MA527192
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fra2 Monoclonal Antibody for IHC (P), Western Blot

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fra2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3
Gene FOSL2
Gene Accession No. P15408
Gene Alias FOS like 2, AP-1 trancription factor subunit; FOS like 2, AP-1 transcription factor subunit; FOS like antigen 2; FOSL2; FOS-like antigen 2; fos-related antigen 2; Fra2; FRA-2
Gene Symbols FOSL2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 110-326 of FOSL2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2355
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.