Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fra2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110687
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110687 100 μL
PIPA5111522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5110687 Supplier Invitrogen™ Supplier No. PA5110687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKLEFMLVAH GPVCKISPEE RRSPPAPGLQ PMRSGGGSVG AVVVKQEPLE EDSPSSSSAG LDKAQRSVIK PISIAGGFYG EEPLHTPIVV TSTPAVTPGT SNLVFTYPSV LEQESPASPS ESCSKAHRRS SSSGDQSSDS L Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 58%.

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fra2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOSL2
Gene Accession No. P15408, P47930, P51145
Gene Alias FOS like 2, AP-1 trancription factor subunit; FOS like 2, AP-1 transcription factor subunit; FOS like antigen 2; FOSL2; FOS-like antigen 2; fos-related antigen 2; Fra2; FRA-2
Gene Symbols FOSL2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Fra2. Recombinant protein control fragment (Product #RP-102469).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14284, 2355, 25446
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.