Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fra2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50 - $596.50
Specifications
Antigen | Fra2 |
---|---|
Concentration | 0.30 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110687
|
Invitrogen™
PA5110687 |
100 μL |
Each for $596.50
|
|
|||||
PIPA5111522
|
Invitrogen™
PA5111522 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: EKLEFMLVAH GPVCKISPEE RRSPPAPGLQ PMRSGGGSVG AVVVKQEPLE EDSPSSSSAG LDKAQRSVIK PISIAGGFYG EEPLHTPIVV TSTPAVTPGT SNLVFTYPSV LEQESPASPS ESCSKAHRRS SSSGDQSSDS L.
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.Specifications
Fra2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
FOSL2 | |
FOS like 2, AP-1 trancription factor subunit; FOS like 2, AP-1 transcription factor subunit; FOS like antigen 2; FOSL2; FOS-like antigen 2; fos-related antigen 2; Fra2; FRA-2 | |
FOSL2 | |
Primary | |
Antigen affinity chromatography |
0.30 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P15408, P47930, P51145 | |
14284, 2355, 25446 | |
Recombinant Protein corresonding to Human Fra2. Recombinant protein control fragment (Product #RP-102469). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title