Learn More
Fracking Model
SDPSCI_ED
Catalog No. S06668
Click to view available options
Material:
Plastic
Shows the controversial method in a simple, noncontroversial way.
Fracking is the most effective new technique to extract natural gas by drilling a horizontal hole at great depth, injecting sand and fluids at high pressure and shattering rock to allow gas to escape.
  • Plastic model shows the basic structure of a modern natural gas well with the relative position of the surface, gas layer and aquifers
  • Provides a unique place to start a discussion on energy
  • Use as a stencil for students to draw their own pictures
Specifications
For Use With (Application) Use as a stencil for students to draw their own pictures
Product Type Fracking Model
Material Plastic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.