Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates
Click to view available options
Surface Treatment:
MaxiSorp
PolySorp
Certified:
No
Yes
Description
Perform quantitative and qualitative solid phase immunoassays with Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates, which include Nunc™, Immulon™ and Microtiter™ plates. Choose from a variety of binding surfaces for reliable and reproducible results in colormetric assays.
96-Well Solid Plates
- Clear polystyrene
- Choice of four surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces
- PolySorp for adsorption of hydrophobic molecules
- MaxiSorp is hydrophilic and ideal for antibody sandwich assays
Ordering Information:
Please note that availability may vary by country. 446612 includes certificate.
Specifications
Specifications
| Barcode | No Barcode |
| Binding Property | Hydrophilic |
| Certified | No |
| Format | Standard |
| Lid | Without Lid |
| Detection Method | Colorimetric |
| Array | 8 x 12 |
| Packaging | 60 Cases |
| Color | Clear |
| Product Type | Microplate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction