Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRAS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562439 Supplier Invitrogen™ Supplier No. PA562439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IWIQLNYLPS YGTLLRISGS EVEELSEVSN FTMEDINNKK IRYSAVFETD GHLVTDSFYF SVSDMDHNHL DNQIFTIMIT PAENPPPVIA FADLITVDEG GRAPLSFHHF FATDDDDNLQ RDAIIKLSAL PKYGCIENTG TGDRFG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.

This gene encodes an extracellular matrix protein that appears to function in the regulation of epidermal-basement membrane adhesion and organogenesis during development. Mutations in this gene cause Fraser syndrome, a multisystem malformation that can include craniofacial, urogenital and respiratory system abnormalities. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRAS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FRAS1
Gene Accession No. Q86XX4
Gene Alias bl; blebbed; E130113P14Rik; extracellular matrix protein FRAS1; FLJ14927; FLJ22031; FRAS1; Fraser extracellular matrix complex subunit 1; Fraser syndrome 1; Fraser syndrome 1 homolog (human); Fraser syndrome 1 protein; Kiaa1500; mKIAA1500
Gene Symbols FRAS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FRAS1. Recombinant protein control fragment (Product #RP-103105).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.