Learn More
3M™ Fraser Broth Base BP0210500, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26825 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Fraser Broth Base BP0210500, Sold by AquaPhoenix Scientific
Catalog No.
S26825 Fraser Broth Base BP0210500, Sold by AquaPhoenix Scientific
1 options
Catalog No. S26825 Supplier 3M™ Supplier No. BP0210500
Add to Cart
Add to Cart
For selective enrichment of Listeria species including Listeria monocytogenes
  • Designed to improve recovery of sublethally damaged cells
  • Easy to use
  • Consistent results
  • Increased plant efficiency
Specifications
Detects Listeria species including Listeria monocytogenes
Product Type Microbiology Media
Test Type Environmental Test
Weight (English) 1.1 lb.
Weight (Metric) 500 g