Learn More
3M™ Fraser Broth Supplement BP0220010, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26826 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
10/Cs.
Catalog No. Quantity
S26826 10/Cs.
1 options
Catalog No. S26826 Supplier 3M™ Supplier No. BP0220010
Add to Cart
Add to Cart
5% ferric ammonium citrate, required for differential enrichment of Listeria
  • Flexibility to make when needed
  • Reduced costs
Specifications
Product Type Microbiology Media
Test Type Environmental Test
For Use With (Application) Differential enrichment of Listeria
Quantity 10/Cs.
Volume (Metric) Specimen 10 mL