Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FRAT2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518023
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
The protein encoded by this intronless gene belongs to the GSK-3-binding protein family. Studies show that this protein plays a role as a positive regulator of the WNT signaling pathway. It may be upregulated in tumor progression.
Specifications
| FRAT2 | |
| Polyclonal | |
| Unconjugated | |
| FRAT2 | |
| FRAT2; FRAT-2; frequently rearranged in advanced T cell lymphomas 2; frequently rearranged in advanced T-cell lymphomas 2; GSK-3; GSK-3 binding protein FRAT2; GSK-3-binding protein FRAT2; MGC10562; RP11-452K12.2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 23401 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O75474 | |
| FRAT2 | |
| Synthetic peptide sequence (GPDRIALQPSGSLL) corresponding to the C-terminus amino acids of FRAT2 (aa 220-233). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction