Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frataxin Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $562.50
Specifications
Antigen | Frataxin |
---|---|
Applications | Western Blot, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Frataxin Polyclonal specifically detects Frataxin in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.Specifications
Frataxin | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 42-210 of human Frataxin (NP_000135.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Diabetes Research | |
PBS with 50% glycerol, pH7.3. | |
2395 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title