Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FRAX 486
GREENER_CHOICESDP

Catalog No. 519010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-901-0 Supplier Tocris Bioscience™ Supplier No. 5190/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent PAK inhibitor; brain penetrant and orally bioavailable

FRAX 486 is a potent p21-activated kinase (PAK) inhibitor (IC50 values are 14, 33, 39 and 575 nM for PAK1, PAK2, PAK3 and PAK4 respectively). Blocks and reverses the DISC1 knockdown-induced reduction in dendritic spine size in cortical neurons. Attenuates dendritic spine elimination and enhances spine generation in DISC1 knockdown mice. Ameliorates autism-like behavioral symptoms in fragile X mental retardation 1 (Fmr1) knockout mice. Brain penetrant and orally bioavailable.

Specifications

Quantity 10 mg
Product Type FRAX 486
Purity >98%
Molecular Weight (g/mol) 513.39
Inhibitors FRAX 486

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.