Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedical™ Free T3 (Triiodothyronine) Coated Tube RIA Kit
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 Tubes
Description
Free T3 (Triiodothyronine) Coated Tube RIA Kit is intended for the measurement of Free T3 in a sample.

Specifications
Specifications
Product Type | Kit and Assay |
Sample Type | Plasma, Serum |
Target Species | Human |
For Use With (Application) | Endocrinology |
Quantity | 100 Tubes |
Regulatory Status | FDA Cleared (K840959) |
Sample Volume | 100 μL |
Storage Requirements | 2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction