Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedical™ Free T4 (Thyroxine) Coated Tube RIA Kit
Click to view available options
Quantity:
500 Tubes
Description
The Free T4 (Thyroxine) Coated Tube RIA Kit is intended for the measurement of Free T4 in a sample.

Specifications
Specifications
Product Type | Kit and Assay |
Sample Type | Plasma, Serum |
Target Species | Human |
For Use With (Application) | Endocrinology |
Host Species | Human |
Quantity | 500 Tubes |
Regulatory Status | FDA Cleared (K823264/A) |
Sample Volume | 50 μL |
Storage Requirements | 2°C to 8°C |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction