Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes Freeze Protected, Pedestal Mount Eye/Face Wash

Catalog No. p-7114654 Shop All Justrite Safety Group Products
Click to view available options
Electrical Requirements:
120 V
240 V
Material (Piping Connection):
Galvanized Steel
Stainless Steel

HughesFreeze Protected, Pedestal Mount Eye/Face Wash is intended for use where there is a possibility of the water freezing due to extreme cold climates and is generally for open areas away from walls.

Features:

  • Trace tape heating with polyethylene jacket and polyurethane foam insulation to improve frost protection and strength.
  • Designed at ANSI-compliant height
  • Suitable for use in both Class 1 Division 2 hazardous areas, as well as non-hazardous areas.
  • Made of durable, corrosion-resistant acrylic capped ABS, the eye/face wash incorporates an integral lid connected to the water valve for automatic operation—activate by simply pulling down the lid.
  • Easy-to-reach flow regulator valve positioned on side of bowl adjusts to suit the severity of the accident.
  • Includes an inspection record tag for recording test and service history.
  • For cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
  • Covered acrylic-capped ABS eye/face wash protects the bowl and eyewash diffusers from dust and debris
  • Diffusers provide a gentle flow of aerated water and prevent further injury to the eyes
  • Includes a separate universal eyewash sign for enhanced visibility
  • Meets ANSI and EN 15154 standards and supports OSHA requirements

Compliance

ANSI Z358.1-2014 and EN15154 Compliant

Warranty and Services

1 Year

Specifications

Activation Pull Down on Lid
Certifications/Compliance ANSI Z358.1-2014 and EN15154 Compliant
Color White with Green Stripes
Depth (English) 24 in.
Depth (Metric) 610 mm
Electrical Requirements 120 V
Flow Rate 3 gpm
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 24 in.
Height (Metric) 610 mm
Includes 1 Unit
Inlet Connection (Metric) 1.25 in. (F) NPT
Material Polyurethane Foam Insulation, Medium Density Impact Resistant Polyethylene, Chrome Plated Brass, Stainless Steel Mesh
Material (Piping Connection) Galvanized Steel
Mounting Pedestal
Pressure 29 to 87 psi
Product Type Safety Shower and Emergency Eye Wash
Width (English) 48 in.
Width (Metric) 1219 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.