Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hughes Freeze Protected, Wall Mounted Drench Shower
Shop All Justrite Safety Group Products

Click to view available options
Electrical Requirements:
120 V
240 V
Material (Piping Connection):
Galvanized Steel
Stainless Steel
Description
Features:
- Trace tape heating with polyethylene jacket and polyurethane foam insulation to improve frost protection and strength
- Ideal for use where space is at a premium
- Specially designed, high-performance quarter-turn ball valve made of 316L stainless steel to endure the harshest environments and site conditions
- Low operating torque to open valve offers reliable, easy on/off operation when emergencies occur
- Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure
- Durable, coated stainless steel shower pull handle with nylon valve connection is easy to grip and activate for immediate water drenching.
- Includes a removable, self-draining overhead nozzle designed for high flow rates that meet or exceed ANSI standards
- Made of durable nylon material, the nozzle offers optimal spray distribution for user protection and water conservation.
- For cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
- Suitable for both Class 1 Division 2 hazardous areas, as well as non-hazardous areas
- Includes a universal safety shower sign for enhanced visibility
- Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements
Compliance
ANSI Z358.1-2014 and EN15154 Compliant
Warranty and Services
1 Year
Specifications
Specifications
Activation | Pull Handle |
Certifications/Compliance | ANSI Z358.1-2014 and EN15154 Compliant |
Color | White with Green Stripes |
Depth (English) | 12 in. |
Depth (Metric) | 305 mm |
Electrical Requirements | 120 V |
For Use With (Application) | Emergency Shower and Eye Wash with Water |
Height (English) | 12 in. |
Height (Metric) | 305 mm |
Includes | 1 Unit |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction