Learn More
Invitrogen™ FREM1 Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-20981 can be used with blocking peptide PEP-1095.
Specifications
Specifications
| Antigen | FREM1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FREM1 |
| Gene Accession No. | Q5H8C1, Q684R7 |
| Gene Alias | BC037594; BNAR; C9orf143; C9orf145; C9orf154; crf11; D430009N09; D630008K06; extracellular matrix protein QBRICK; eye>m02Jus>; eyes2; Fras1 related extracellular matrix 1; Fras1 related extracellular matrix protein 1; FRAS1-related extracellular matrix protein 1; Frem1; heb; MOTA; Protein QBRICK; qbrick; RGD1306981; RP11-265B7.2; TILRR; TRIGNO2 |
| Gene Symbols | FREM1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.