Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FREM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120413 Supplier Invitrogen™ Supplier No. PA5120413
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, HepG2, 293T. The target is usually found in the following locations: Cell membrane, Extracellular side, Single-pass type I membrane protein. Immunogen sequence: QPAAFGRALL SPGLAGAAGV PAEEAIVLAN RGLRVPFGRE VWLDPLHDLV LQVQPGDRCA VSVLDNDALA QRPGRLSPKR FPCDFGPGEV RYSHLGARSP SRDRVRLQLR YDAPGGAVVL PLVLEVEVVF TQLEVVTRNL PLVVEELLGT SNALDARSLE FAFQPETEEC RVGILSGLGA LPRYGELLHY PQVPGGAREG GAPETLLMDC KAFQELGVRY RHTAASRSPN RDWIPMVVEL RSRGAPVGSP ALKR.

FREM2 is a member of the FRAS1-related extracellular matrix protein family and like FREM1, mutations in the FREM2 gene are associated with Fraser syndrome. FREM2 localizes to the basement membrane, forming a ternary complex that plays a role in epidermal-dermal interactions during morphogenetic processes and is thought to be required for maintaining the integrity of the skin epithelium and the differentiated state of renal epithelia. The FREM2 gene is one of several genes whose transcription is affecteded by TFAP2C, a transcription factor involved in mammary development, differentiation, and oncogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FREM2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FREM2
Gene Accession No. Q5SZK8, Q6NVD0
Gene Alias 6030440P17Rik; 8430406N05Rik; b2b1562Clo; ECM3 homolog; FRAS1 related extracellular matrix protein 2; FRAS1-related extracellular matrix protein 2; FREM2; Gm409; my; ne; NV domain containing protein NV1; NV domain-containing protein 1; Nv1
Gene Symbols FREM2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242022, 341640
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.