Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frequenin Antibody (11B8), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259480G
This item is not returnable.
View return policy
Description
Frequenin Monoclonal antibody specifically detects Frequenin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
Frequenin | |
Monoclonal | |
DyLight 488 | |
DKFZp761L1223, FREQFLUP, frequenin (Drosophila) homolog, Frequenin homolog, frequenin homolog (Drosophila), Frequenin-like protein, Frequenin-like ubiquitous protein, NCS-1, neuronal calcium sensor 1 | |
Recombinant human Frequenin (1-190aa) purified from E. coli | |
0.1 mL | |
Neuroscience | |
23413 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA | |
11B8 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction