Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frequenin Mouse anti-Human, Clone: 11B8, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | Frequenin |
---|---|
Clone | 11B8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25948050
![]() |
Novus Biologicals
NBP25948050UL |
50 μL |
Each for $340.50
|
|
|||||
NBP259480X
![]() |
Novus Biologicals
NBP259480100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
Frequenin Monoclonal specifically detects Frequenin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Frequenin | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
DKFZp761L1223, FREQFLUP, frequenin (Drosophila) homolog, Frequenin homolog, frequenin homolog (Drosophila), Frequenin-like protein, Frequenin-like ubiquitous protein, NCS-1, neuronal calcium sensor 1 | |
NCS1 | |
IgG1 κ | |
Protein A purified |
11B8 | |
Western Blot 1:1000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
23413 | |
Recombinant human NCS1 (1-190aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title