Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Freshwater Mussels
SDPSCI_ED

For use in exploring bivalve anatomy. Carolina™ Freshwater Mussels belongs to Unio or Anodonta sp.

$2.54

Specifications

Injection Type Plain
Preservative Formalin
Type Preserved Specimen
Packaging Pail
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
S37689
SDPSCI_ED
View Documents
Carolina™
224846
3 to 4 in. 7.6 to 10.1 cm
Each for $2.54
Add to cart
 
S37688
SDPSCI_ED
View Documents
Carolina™
S224844
4 to 5 in. 10.1 to 12.7 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Freshwater mollusks
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
Specifications

Specifications

Plain
Formalin
Preserved Specimen
Pail
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit