Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vileda Professional™ Evolution Telescopic Handle
Click to view available options
Material:
Aluminum
Description
- Constructed of aluminum
- Extremely lightweight with ergonomic hand grip at the top
- Power grip collars for easy opening/closing even with wet hands
- O-Rings on cone provides extra stability
Specifications
Specifications
Product Type | Telescopic Handle |
Color | Silver, Red |
Length (English) | 78 to 186 in. |
Includes | Ergonomic Center and Anti-slip Grips |
For Use With (Equipment) | Swep Duo Plus Frame |
For Use With (Application) | Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs |
Material | Aluminum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction