Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vileda Professional™ PVAmicro Cloth
Shop All Vileda Professional Products
Click to view available options
Color:
Blue
Green
Red
Description
- Knitted microfiber ensures perfect results on almost all surfaces: glass, labs, stainless steel and more
- The PVA as very good water absorbency, meaning streak-free wiping without after-drying
- PVAmicro™ holds 40 times less particle residue after rinsing, compared to knitted microfiber cloth
- Streak and scratch free performance
Specifications
Specifications
| Product Type | PVAmicro Cloth |
| Color | Blue |
| Dimensions (L x W) | 38 x 35 cm |
| For Use With (Application) | For cleaning all types of hard surfaces |
| Packaging Quantity | 5/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction