Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRG1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299204F
Description
FRG1 Polyclonal specifically detects FRG1 in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
FRG1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
facioscapulohumeral muscular dystrophy region gene-1, FSG1FRG1A, FSHD region gene 1, FSHD region gene 1 protein, protein FRG1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of mouse FRG1. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
2483 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction