Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116240
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116240 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116240 Supplier Invitrogen™ Supplier No. PA5116240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FRG1 Polyclonal Antibody for Western Blot

This gene maps to a location 100 kb centromeric of the repeat units on chromosome 4q35 which are deleted in facioscapulohumeral muscular dystrophy . It is evolutionarily conserved and has related sequences on multiple human chromosomes but DNA sequence analysis did not reveal any homology to known genes. In vivo studies demonstrate the encoded protein is localized to the nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FRG1
Gene Accession No. Q14331
Gene Alias facioscapulohumeral muscular dystrophy region gene-1; Frg1; FRG1A; Frg1l1; FSG1; FSHD region gene 1; FSHD region gene 1 protein; FSHD region gene 1-like 1; hypothetical protein LOC550491; protein FRG1; Unknown (protein for MGC:140313); zgc:112426
Gene Symbols FRG1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 230-256 amino acids from the C-terminal region of human FRG1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.